Thermo Scientific Chemicals

4-n-Decylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A17234.14
25 g, Each
Thermo Scientific Chemicals

4-n-Decylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A17234.14
25 g, Each
Quantity
Catalog number: A17234.14
also known as A17234-14
Price (USD)
Price: 608.00
Online price: 525.65
Your price:
Quantity
-

Chemical Identifiers

CAS
54256-43-8
IUPAC Name
4-decylbenzoyl chloride
Molecular Formula
C17H25ClO
InChI Key
HEWLDHSFOQXCSI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.5115-1.5155 @ 20?C
Form
Liquid

Description

Employed as a pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical and chemical intermediate.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text