Thermo Scientific Chemicals

Anthracene, 97%, Thermo Scientific Chemicals

Catalog number: A17261.0B
1000 g, Each
Thermo Scientific Chemicals

Anthracene, 97%, Thermo Scientific Chemicals

Catalog number: A17261.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A17261.0B
also known as A17261-0B
Price (USD)
Price: 333.00
Your price:
Quantity
-

Chemical Identifiers

CAS
120-12-7
IUPAC Name
anthracene
Molecular Formula
C14H10
InChI Key
MWPLVEDNUUSJAV-UHFFFAOYSA-N
SMILES
C1=CC2=CC3=CC=CC=C3C=C2C=C1
Appearance (Color)
Pale cream to cream to yellow to green to pale brown to black
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
213.0-222.0?C

Description

Anthracene is used as a source of dyestuffs and involved in the preparation of red dye alizarin. It is an organic semiconductor used as a scintillator for detectors of high energy photons, electrons and alfa particles. It is doped with polyvinyltoluene to prepare plastic scintillator. Further, it is used in the preparation of anthraquinone, wood preservatives, coating materials and pigments. In addition to this, it is used to prepare 1-hydroxyanthracene, 2-hydroxyanthracene and in 9,10-dihydroxyanthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene is used as a source of dyestuffs and involved in the preparation of red dye alizarin. It is an organic semiconductor used as a scintillator for detectors of high energy photons, electrons and alfa particles. It is doped with polyvinyltoluene to prepare plastic scintillator. Further, it is used in the preparation of anthraquinone, wood preservatives, coating materials and pigments. In addition to this, it is used to prepare 1-hydroxyanthracene, 2-hydroxyanthracene and in 9,10-dihydroxyanthracene.

Solubility
Soluble in methanol, benzene, chloroform, ether, carbon disulfide, carbon tetrachloride and toluene. Slightly soluble in acetone. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and hypochlorites.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text