2-Chloroethyl isocyanate, 97%
2-Chloroethyl isocyanate, 97%
2-Chloroethyl isocyanate, 97%
Thermo Scientific Chemicals

2-Chloroethyl isocyanate, 97%

CAS: 1943-83-5 | C3H4ClNO | 105.521 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A17289.06
または、製品番号A17289-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1943-83-5
IUPAC Name1-chloro-2-isocyanatoethane
Molecular FormulaC3H4ClNO
InChI KeyBCMYXYHEMGPZJN-UHFFFAOYSA-N
SMILESClCCN=C=O
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4435-1.4485 @ 20°C (non-U.S. specification)
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Hydrolyzes in water.

Notes
Store at 4°C. Incompatible with alcohols. Amines, oxidizing agents. Hygroscopic.
RUO – Research Use Only

General References:

  1. John R. Babson. ; Donald J. Reed. Inactivation of glutathione reductase by 2-chloroethyl nitrosourea-derived isocyanates.Biochem. Biophys. Res. Commun.. 1978, 83 (2),754-762.
  2. John A. Montgomery.; Ruby. James.; George S. McCaleb.; Thomas P. Johnston. The Modes of Decomposition of 1,3-Bis(2-chloroethyl)-1-nitrosourea and Related Compounds. J. Med. Chem. 1967, 10 (4),668-674.