Isopropyl trifluoroacetate, 97%
Isopropyl trifluoroacetate, 97%
Isopropyl trifluoroacetate, 97%
Isopropyl trifluoroacetate, 97%
Thermo Scientific Chemicals

Isopropyl trifluoroacetate, 97%

CAS: 400-38-4 | C5H7F3O2 | 156.10 g/mol
製品番号(カタログ番号) A17322.14
または、製品番号A17322-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS400-38-4
IUPAC Namepropan-2-yl 2,2,2-trifluoroacetate
Molecular FormulaC5H7F3O2
InChI KeyASAXRKSDVDALDT-UHFFFAOYSA-N
SMILESCC(C)OC(=O)C(F)(F)F
さらに表示
Appearance (Color)Clear colorless to pale yellow or pink
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3165-1.3215 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only