Thermo Scientific Chemicals

Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals

Catalog number: A17361.0B
1000 g, Each
Thermo Scientific Chemicals

Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals

Catalog number: A17361.0B
1000 g, Each
Quantity
Catalog number: A17361.0B
also known as A17361-0B
Price (USD)
Price: 194.00
Online price: 164.65
Your price:
Quantity
-

Chemical Identifiers

CAS
39339-85-0
MDL Number
MFCD00145579
Comment
Exchange capacity: ≥0.80 eq/L
Particle Size
0.56-0.70mm
Form
Beads
Appearance (Color)
Moist pink or pale brown
Comment
Moisture holding capacity: 66-75% (OH form)

Description

Amberlyst™ A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlyst|r A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

Solubility
Insoluble in water.

Notes
Exposure to air absorbs carbon dioxide. Incompatible with strong acids, bases and oxidants.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text