(-)-Diisopropyl D-tartrate, 98%
(-)-Diisopropyl D-tartrate, 98%
(-)-Diisopropyl D-tartrate, 98%
Thermo Scientific Chemicals

(-)-Diisopropyl D-tartrate, 98%

CAS: 62961-64-2 | C10H18O6 | 234.248 g/mol
製品番号(カタログ番号) A17362.14
または、製品番号A17362-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS62961-64-2
IUPAC Name1,4-bis(propan-2-yl) (2R,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC10H18O6
InChI KeyXEBCWEDRGPSHQH-OCAPTIKFSA-N
SMILESCC(C)OC(=O)[C@@H](O)[C@@H](O)C(=O)OC(C)C
さらに表示
Appearance (Color)Clear colorless
Optical Rotation-15.5 ± 1.0? (neat)
Assay (GC)≥97.5%
Refractive Index1.4375-1.4415 @ 20?C
FormLiquid or viscous liquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only