Tetramethylammonium hexafluorophosphate, 99%
Tetramethylammonium hexafluorophosphate, 99%
Tetramethylammonium hexafluorophosphate, 99%
Thermo Scientific Chemicals

Tetramethylammonium hexafluorophosphate, 99%

CAS: 558-32-7 | C4H12F6NP | 219.111 g/mol
製品番号(カタログ番号) A17467.14
または、製品番号A17467-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS558-32-7
IUPAC Namehexafluoro-λ⁵-phosphanuide; tetramethylazanium
Molecular FormulaC4H12F6NP
InChI KeyZFKULDQWLXAKIM-UHFFFAOYSA-N
SMILESC[N+](C)(C)C.F[P-](F)(F)(F)(F)F
さらに表示
Appearance (Color)White to pale cream
Elemental AnalysisHydrogen H: 5.44-5.60% (Theory 5.52%)
Melting Point>300?C
Elemental AnalysisCarbon C: 21.60-22.25% (Theory 21.93%)
Assay (unspecified)≥98.5% by CHN analysis
さらに表示
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only

General References:

  1. Y. Wang, et al.The structure of tetramethylammonium hexafluorophosphate.Acta Cryst.,1980,B361523.
  2. Albert J. Fry, et al. Electrolyte effects upon the polarographic reduction of alkyl halides in dimethyl sulfoxide.J. Org. Chem.,1976,41(1), 54-57.