Ammonium magnesium phosphate hexahydrate, 98%
Ammonium magnesium phosphate hexahydrate, 98%
Ammonium magnesium phosphate hexahydrate, 98%
Thermo Scientific Chemicals

Ammonium magnesium phosphate hexahydrate, 98%

CAS: 13478-16-5 | H16MgNO10P | 245.404 g/mol
製品番号(カタログ番号) A17497.36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13478-16-5
IUPAC Namemagnesium(2+) ammonium hexahydrate phosphate
Molecular FormulaH16MgNO10P
InChI KeyCKMXBZGNNVIXHC-UHFFFAOYSA-L
SMILES[NH4+].O.O.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
さらに表示
Loss on Drying45.0-50.0% (200?C/constant wt.)
Total Metal ImpuritiesIron Fe: ≤20ppm
FormPowder
Identification (FTIR)Conforms
Total Metal ImpuritiesCobalt Co: ≤10ppm
さらに表示
It is used as pharmaceutical intermediae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediae.

Solubility
Insoluble in water.

Notes
Store in a cool place. Ensure proper ventilation.
RUO – Research Use Only

General References:

  1. Nora E Hipedinger.; Alberto N Scian.; Esteban F Aglietti. Magnesia-ammonium phosphate-bonded cordierite refractory castables: Phase evolution on heating and mechanical properties. Cem. Concr. Res. 2004, 34 (1),157-164.
  2. Henry Bassett.; William L. Bedwell. Studies of phosphates. Part I. Ammonium magnesium phosphate and related compounds .J. Chem. Soc. 1933, (0),854-871.