1-Tetradecylphosphonic acid, 98%
1-Tetradecylphosphonic acid, 98%
1-Tetradecylphosphonic acid, 98%
Thermo Scientific Chemicals

1-Tetradecylphosphonic acid, 98%

CAS: 4671-75-4 | C14H31O3P | 278.373 g/mol
製品番号(カタログ番号) A17511.06
または、製品番号A17511-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4671-75-4
IUPAC Nametetradecylphosphonic acid
Molecular FormulaC14H31O3P
InChI KeyBVQJQTMSTANITJ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCP(O)(O)=O
さらに表示
Melting Point (clear melt)94.0-101.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ye, H. J.; Shao, W. Z.; Zhen, L. Tetradecylphosphonic acid modified BaTiO3 nanoparticles and its nanocomposite. Colloids Surf., A 2013, 427, 19-25.
  2. Wang, H.-r.; Jiang, Y.; Qu, J.-e.; Cao, Z.-y.; Liu, S.-b.; Guo, X.-p. Adsorption and Corrosion Inhibition Performances of 1-Tetradecylphosphonic Acid Self-Assembled Monolayers on 430 Stainless Steel Surface. J. Iron. Steel Res. Int. 2013, 20 (9), 93-98.