Thermo Scientific™

Isosorbide dimethyl ether, 98%, Thermo Scientific Chemicals

Catalog number: A17516.14
25 g, Each
Thermo Scientific™

Isosorbide dimethyl ether, 98%, Thermo Scientific Chemicals

Catalog number: A17516.14
25 g, Each
Quantity
Catalog number: A17516.14
also known as A17516-14
Price (USD)
Price: 63.40
Online price: 54.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5306-85-4
IUPAC Name
3,6-dimethoxy-hexahydrofuro[3,2-b]furan
Molecular Formula
C8H14O4
InChI Key
MEJYDZQQVZJMPP-UHFFFAOYNA-N
SMILES
COC1COC2C(COC12)OC
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4605-1.4645 @ 20?C

Description

It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.

Solubility
Soluble in water.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text