2-Hydroxy-4-methoxybenzophenone, 98+%
2-Hydroxy-4-methoxybenzophenone, 98+%
2-Hydroxy-4-methoxybenzophenone, 98+%
2-Hydroxy-4-methoxybenzophenone, 98+%
Thermo Scientific Chemicals

2-Hydroxy-4-methoxybenzophenone, 98+%

CAS: 131-57-7 | C14H12O3 | 228.247 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A17662.30
または、製品番号A17662-30
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS131-57-7
IUPAC Name2-benzoyl-5-methoxyphenol
Molecular FormulaC14H12O3
InChI KeyDXGLGDHPHMLXJC-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
さらに表示
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to yellow
Assay (GC)≥98.0%
Melting Point (clear melt)60.0-67.0?C
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.

Solubility
Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

General References:

  1. Yiin, L. M.; Tian, J. N.; Hung, C. C. Assessment of dermal absorption of DEET-containing insect repellent and oxybenzone-containing sunscreen using human urinary metabolites. Environ. Sci. Pollut. Res. 2015, 22 (9), 7062-7070.
  2. Louis, G. M. B.; Chen, Z.; Kim, S.; Sapra, K. J.; Bae, J.; Kannan, K. Urinary concentrations of benzophenone-type ultraviolet light filters and semen quality. Fertil. Steril. 2015, 104 (4), 989-996.