3,4-Dichlorobenzyl chloride, 98%
3,4-Dichlorobenzyl chloride, 98%
3,4-Dichlorobenzyl chloride, 98%
3,4-Dichlorobenzyl chloride, 98%
Thermo Scientific Chemicals

3,4-Dichlorobenzyl chloride, 98%

CAS: 102-47-6 | C7H5Cl3 | 195.467 g/mol
数量:
25 g
50 g
100 g
250 g
製品番号(カタログ番号) A17672.14
または、製品番号A17672-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS102-47-6
IUPAC Name1,2-dichloro-4-(chloromethyl)benzene
Molecular FormulaC7H5Cl3
InChI KeyYZIFVWOCPGPNHB-UHFFFAOYSA-N
SMILESClCC1=CC=C(Cl)C(Cl)=C1
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5755-1.5795 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only