2-Chlorobenzothiazole, 98%
2-Chlorobenzothiazole, 98%
2-Chlorobenzothiazole, 98%
Thermo Scientific Chemicals

2-Chlorobenzothiazole, 98%

CAS: 615-20-3 | C7H4ClNS | 169.626 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A17762.22
または、製品番号A17762-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS615-20-3
IUPAC Name2-chloro-1,3-benzothiazole
Molecular FormulaC7H4ClNS
InChI KeyBSQLQMLFTHJVKS-UHFFFAOYSA-N
SMILESClC1=NC2=CC=CC=C2S1
さらに表示
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.6360-1.6400 @ 20?C
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only