N-Methylanilinium trifluoroacetate, 99%
N-Methylanilinium trifluoroacetate, 99%
N-Methylanilinium trifluoroacetate, 99%
Thermo Scientific Chemicals

N-Methylanilinium trifluoroacetate, 99%

CAS: 29885-95-8 | C7H10N·CF3CO2
製品番号(カタログ番号) A17781.22
または、製品番号A17781-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS29885-95-8
IUPAC NameN-methylanilinium trifluoroacetate
Molecular FormulaC9H10F3NO2
InChI KeyTXXJGCVOHDSYBC-UHFFFAOYSA-N
SMILES[O-]C(=O)C(F)(F)F.C[NH2+]C1=CC=CC=C1
さらに表示
Appearance (Color)White to brown to gray
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)62.0-68.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only