N,N-Diethyl-p-phenylenediamine sulfate, 97%
N,N-Diethyl-p-phenylenediamine sulfate, 97%
N,N-Diethyl-p-phenylenediamine sulfate, 97%
Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine sulfate, 97%

CAS: 6283-63-2 | C10H18N2O4S | 262.324 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A17832.30
または、製品番号A17832-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6283-63-2
IUPAC NameN1,N1-diethylbenzene-1,4-diamine; sulfuric acid
Molecular FormulaC10H18N2O4S
InChI KeyAYLDJQABCMPYEN-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CCN(CC)C1=CC=C(N)C=C1
さらに表示
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)182-189?C
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Pachamuthu, M. P.; Karthikeyan, S.; Sekaran, G.; Maheswari, R.; Ramanathan, A. Fenton-Type Oxidative Degradation of N,N-Diethyl-p-phenylenediamine by a Mesoporous Wormhole Structured FeTUD-1 Catalyst. Clean: Soil, Air, Water 2015, 43 (3), 375-381.
  2. Melnikova, L.; Petrenko, V. I.; Avdeev, M. V.; Ivankov, O. I.; Bulavin, L. A.; Garamus, V. M.; Almásy, L.; Mitroova, Z.; Kopcansky, P. SANS contrast variation study of magnetoferritin structure at various iron loading. J. Magn. Magn. Mater. 2015, 377, 77-80.
  3. Criado, P.; Fraschini, C.; Salmieri, S.; Becher, D.; Safrany, A.; Lacroix, M. Evaluation of Antioxidant Cellulose Nanocrystals and Applications in Gellan Gum Films. Ind. Biotechnol. 2015, 11 (1), 59-68.