Thermo Scientific Chemicals

Methyl glycolate, 98%, Thermo Scientific Chemicals

Catalog number: A17870.14
25 g, Each
Thermo Scientific Chemicals

Methyl glycolate, 98%, Thermo Scientific Chemicals

Catalog number: A17870.14
25 g, Each
Quantity
Catalog number: A17870.14
also known as A17870-14
Price (USD)
Price: 116.00
Online price: 99.65
Your price:
Quantity
-

Chemical Identifiers

CAS
96-35-5
IUPAC Name
methyl 2-hydroxyacetate
Molecular Formula
C3H6O3
InChI Key
GSJFXBNYJCXDGI-UHFFFAOYSA-N
SMILES
COC(=O)CO
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤1%

Description

Methyl glycolate is a chemical reagent used in the synthesis of taxol and rapamycin. It is used in leather tanning and textile dyeing. It acts as a flavoring agent, preservative in food processing, in water treatment industries and as chemical intermediates. Further, it is used in emulsion polymers, ink and paint additive to improve flow properties and impart gloss. It plays an important role in producing triisopropylsilanyloxy-acetic acid methyl ester using imidazole as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl glycolate is a chemical reagent used in the synthesis of taxol and rapamycin. It is used in leather tanning and textile dyeing. It acts as a flavoring agent, preservative in food processing, in water treatment industries and as chemical intermediates. Further, it is used in emulsion polymers, ink and paint additive to improve flow properties and impart gloss. It plays an important role in producing triisopropylsilanyloxy-acetic acid methyl ester using imidazole as reagent.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text