Thermo Scientific Chemicals

(1-Dodecyl)trimethylammonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: A17924.14
25 g, Each
Thermo Scientific Chemicals

(1-Dodecyl)trimethylammonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: A17924.14
25 g, Each
Quantity
Catalog number: A17924.14
also known as A17924-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
112-00-5
IUPAC Name
dodecyltrimethylazanium chloride
Molecular Formula
C15H34ClN
InChI Key
DDXLVDQZPFLQMZ-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCCCCCC[N+](C)(C)C
Infrared spectrum
Conforms
Appearance (Color)
White to pale yellow
Assay from Suppliers CofA
≥96.0%
Form
Powder or crystals or crystalline powder

Description

Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text