1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals
1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals
1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals
1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
A17953.18
または、製品番号A17953-18
50 g
製品番号(カタログ番号) A17953.18
または、製品番号A17953-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1435-48-9
IUPAC Name2,4-dichloro-1-fluorobenzene
Molecular FormulaC6H3Cl2F
InChI KeyBDJZCCWUSOZUQG-UHFFFAOYSA-N
SMILESFC1=CC=C(Cl)C=C1Cl
さらに表示
Refractive Index1.5240-1.5260 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only