Thermo Scientific Chemicals

Cadmium iodide, 98%, Thermo Scientific Chemicals

Catalog number: A18079.22
100 g, Each
Thermo Scientific Chemicals

Cadmium iodide, 98%, Thermo Scientific Chemicals

Catalog number: A18079.22
100 g, Each
Quantity
Catalog number: A18079.22
also known as A18079-22
Price (USD)

Chemical Identifiers

CAS
7790-80-9
IUPAC Name
cadmium(2+) diiodide
Molecular Formula
CdI2
InChI Key
OKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES
[Cd++].[I-].[I-]
Appearance (Color)
White to cream or pale yellow
Form
Powder or crystalline powder or crystals and/or flakes
Complexometric Titration
≥97.5 to ≤102.5%

Description

It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.

Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.

Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text