4-Phenylbutyric acid, 99%
4-Phenylbutyric acid, 99%
4-Phenylbutyric acid, 99%
4-Phenylbutyric acid, 99%
Thermo Scientific Chemicals

4-Phenylbutyric acid, 99%

CAS: 1821-12-1 | C10H12O2 | 164.204 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A18115.36
または、製品番号A18115-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1821-12-1
IUPAC Name4-phenylbutanoic acid
Molecular FormulaC10H12O2
InChI KeyOBKXEAXTFZPCHS-UHFFFAOYSA-N
SMILESOC(=O)CCCC1=CC=CC=C1
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or chunks or fused solid
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (GC)≥98.5%
4-Phenylbutyric acid is used as a chemical chaperone involved in protein-folding disorders. It is involved in the synthesis of dyes and an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylbutyric acid is used as a chemical chaperone involved in protein-folding disorders. It is involved in the synthesis of dyes and an active pharmaceutical ingredient intermediate.

Solubility
Soluble in water and methanol.

Notes
Styrene-DVB; Strongly basic gel type resin.Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kolb, P. S.; Ayaub, E. A.; Zhou, W.; Yum, V.; Dickhout, J. G.; Ask, K. The therapeutic effects of 4-phenylbutyric acid in maintaining proteostasis. Int. J. Biochem. Cell Biol. 2015, 61, 45-52.
  2. Hachuła, B.; Jabłońska-Czapla, M.; Flakus, H. T.; Nowak, M.; Kusz, J. H/D isotopic recognition and temperature effects in IR spectra of hydrogen-bonded cyclic dimers in crystals: 3-Methylcinnamic acid and 4-phenylbutyric acid. Spectrochim. Acta, Part A 2015, 134, 592-597.