1,4-Dibromotetrafluorobenzene, 99%
1,4-Dibromotetrafluorobenzene, 99%
1,4-Dibromotetrafluorobenzene, 99%
1,4-Dibromotetrafluorobenzene, 99%
Thermo Scientific Chemicals

1,4-Dibromotetrafluorobenzene, 99%

CAS: 344-03-6 | C6Br2F4 | 307.87 g/mol
製品番号(カタログ番号) A18135.03
または、製品番号A18135-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS344-03-6
IUPAC Name1,4-dibromo-2,3,5,6-tetrafluorobenzene
Molecular FormulaC6Br2F4
InChI KeyQFTZULJNRAHOIY-UHFFFAOYSA-N
SMILESFC1=C(F)C(Br)=C(F)C(F)=C1Br
さらに表示
Appearance (Color)White to cream
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or chunks
Melting Point (clear melt)75.0-82.0?C
It is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chemical intermediate.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. A. Keith. Jameson.; Cynthia J. Jameson.Absolute temperature dependence of chemical shifts of lock solvents. Tetramethylsilane, hexafluorobenzene, and 1,4-dibromotetrafluorobenzene. Am. Chem. Soc.1973,95(26), 8559-8561.
  2. Alessandra Forni. Experimental and Theoretical Study of the Br···N Halogen Bond in Complexes of 1,4-Dibromotetrafluorobenzene with Dipyridyl Derivatives.J. Phys. Chem. A,2009,113(14), 3403-3412.