p-Xylylenebis(triphenylphosphonium bromide), 96%
p-Xylylenebis(triphenylphosphonium bromide), 96%
p-Xylylenebis(triphenylphosphonium bromide), 96%
Thermo Scientific Chemicals

p-Xylylenebis(triphenylphosphonium bromide), 96%

CAS: 40817-03-6 | C44H38Br2P2 | 788.54 g/mol
製品番号(カタログ番号) A18241.14
または、製品番号A18241-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS40817-03-6
IUPAC Nametriphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dibromide
Molecular FormulaC44H38Br2P2
InChI KeyZZQVVCXWFPGKJD-UHFFFAOYSA-L
SMILES[Br-].[Br-].C(C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
FormPowder
Appearance (Color)White
Loss on Drying≤3% (105?C/constant weight)
Assay (Titration ex Bromide)≥95.0 to ≤105.0%
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Babu Rao Renikuntla; Heather C.Rose; Joby Eldo; Alan S.Waggoner; Bruce A.Armitage. Improved photostability and fluorescence properties through polyfluorination of a cyanine dye. Org. Lett. 2004, 6,(6), 909-912.
  2. Michelle L.Renak ; Glenn P.Bartholomew; Shujun Wang; Pascal J.Ricatto; Rene J.Lachicotte; Guillermo C.Bazan. Fluorinated distyrylbenzene chromophores: Effect of fluorine regiochemistry on molecular properties and solid-state organization. J. Am. Chem. Soc. 1999, 121,(34), 7787-7799.