1-Naphthyl phosphate monosodium salt monohydrate, 98+%
1-Naphthyl phosphate monosodium salt monohydrate, 98+%
1-Naphthyl phosphate monosodium salt monohydrate, 98+%
Thermo Scientific Chemicals

1-Naphthyl phosphate monosodium salt monohydrate, 98+%

CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A18254.22
または、製品番号A18254-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS81012-89-7
Water Content (Karl Fischer Titration)3.53-9.89% (0.5-1.5 waters)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Substrate for phosphatase: J. Biol. Chem., 7, 190 (1951).
  2. Renglin, A.; Olsson, A.; Wachtmeister, C. A.; Onfelt, A. Mitotic disturbance by carbaryl and the metabolite 1-naphthol may involve kinase-mediated phosphorylation of 1-naphthol to the protein phosphatase inhibitor 1-naphthylphosphate. Mutagenesis. 1998, 13 (4), 345-352.
  3. Bowers, J. L.; Bowers Jr, G. N. 1-naphthyl phosphate: specifications for high-quality substrate for measuring prostatic acid phosphatase activity. Clin. Chem. 1982, 28 (1), 212-215.