4-Nitroveratrole, 98+%
4-Nitroveratrole, 98+%
4-Nitroveratrole, 98+%
Thermo Scientific Chemicals

4-Nitroveratrole, 98+%

CAS: 709-09-1 | C8H9NO4 | 183.16 g/mol
数量:
25 g
50 g
250 g
製品番号(カタログ番号) A18264.14
または、製品番号A18264-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS709-09-1
IUPAC Name1,2-dimethoxy-4-nitrobenzene
Molecular FormulaC8H9NO4
InChI KeyYFWBUVZWCBFSQN-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)[N+]([O-])=O
さらに表示
Melting Point (clear melt)94.5-100.5?C
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only