N,N-Dimethyl-4-nitrosoaniline, 98%
N,N-Dimethyl-4-nitrosoaniline, 98%
N,N-Dimethyl-4-nitrosoaniline, 98%
Thermo Scientific Chemicals

N,N-Dimethyl-4-nitrosoaniline, 98%

CAS: 138-89-6 | C8H10N2O | 150.181 g/mol
数量:
25 g
100 g
250 g
製品番号(カタログ番号) A18301.14
または、製品番号A18301-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS138-89-6
IUPAC NameN,N-dimethyl-4-nitrosoaniline
Molecular FormulaC8H10N2O
InChI KeyCMEWLCATCRTSGF-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=O
さらに表示
Appearance (Color)Green
FormCrystals or powder
Assay (GC)>97.5%
Solution Test1% w/v in ethanol : Clear
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only