3-Nitro-5-(trifluoromethyl)benzoic acid, 97%
3-Nitro-5-(trifluoromethyl)benzoic acid, 97%
3-Nitro-5-(trifluoromethyl)benzoic acid, 97%
Thermo Scientific Chemicals

3-Nitro-5-(trifluoromethyl)benzoic acid, 97%

CAS: 328-80-3 | C8H4F3NO4 | 235.118 g/mol
製品番号(カタログ番号) A18316.06
または、製品番号A18316-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS328-80-3
IUPAC Name3-nitro-5-(trifluoromethyl)benzoate
Molecular FormulaC8H3F3NO4
InChI KeyODCLHXGXGFBBTA-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC(=CC(=C1)C(F)(F)F)[N+]([O-])=O
さらに表示
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Appearance (Color)Pale cream to pale yellow
Melting Point (clear melt)125.0-135.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only