Thermo Scientific Chemicals

Nitrilotriacetic acid trisodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A18348.22
100 g, Each
Thermo Scientific Chemicals

Nitrilotriacetic acid trisodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A18348.22
100 g, Each
Quantity
Catalog number: A18348.22
also known as A18348-22
Price (USD)
Price: 20.10
Online price: 17.65
Your price:
Quantity
-

Chemical Identifiers

CAS
18662-53-8
IUPAC Name
trisodium 2-[bis(carboxylatomethyl)amino]acetate hydrate
Molecular Formula
C6H8NNa3O7
InChI Key
HERBOKBJKVUALN-UHFFFAOYSA-K
SMILES
O.[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)CC([O-])=O
Appearance (Color)
White
Assay (ex Sulphated Ash)
≥98.0%
Form
Crystals or powder or crystalline powder or fused solid
Water Content (Karl Fischer Titration)
3.38-9.51% (0.5-1.5 waters)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text