2-Mercaptobenzimidazole, 97%
2-Mercaptobenzimidazole, 97%
2-Mercaptobenzimidazole, 97%
Thermo Scientific Chemicals

2-Mercaptobenzimidazole, 97%

CAS: 583-39-1 | C7H6N2S | 150.20 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A18350.0E
または、製品番号A18350-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS583-39-1
IUPAC Name2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular FormulaC7H6N2S
InChI KeyYHMYGUUIMTVXNW-UHFFFAOYSA-N
SMILESS=C1NC2=CC=CC=C2N1
さらに表示
Appearance (Color)White to pale brown
FormPowder or crystalline powder
Assay (HPLC)≥96.0%
2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Zhong, B.; Jia, Z.; Luo, Y.; Guo, B.; Jia, D. Preparation of halloysite nanotubes supported 2-mercaptobenzimidazole and its application in natural rubber. Composites Part A 2015, 73, 63-71.
  2. Žerjav, G.; Milošev, I. Protection of copper against corrosion in simulated urban rain by the combined action of benzotriazole, 2-mercaptobenzimidazole and stearic acid. Corros. Sci. 2015, 98, 180-191.