3,5-Dibromo-4-hydroxybenzoic acid, 98%
3,5-Dibromo-4-hydroxybenzoic acid, 98%
3,5-Dibromo-4-hydroxybenzoic acid, 98%
Thermo Scientific Chemicals

3,5-Dibromo-4-hydroxybenzoic acid, 98%

CAS: 3337-62-0 | C7H4Br2O3 | 295.914 g/mol
製品番号(カタログ番号) A18361.14
または、製品番号A18361-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3337-62-0
IUPAC Name3,5-dibromo-4-hydroxybenzoic acid
Molecular FormulaC7H4Br2O3
InChI KeyPHWAJJWKNLWZGJ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Br)=C(O)C(Br)=C1
さらに表示
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Assay (Silylated GC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only