Succinyl chloride, 96%
Succinyl chloride, 96%
Succinyl chloride, 96%
Thermo Scientific Chemicals

Succinyl chloride, 96%

CAS: 543-20-4 | C4H4Cl2O2 | 154.974 g/mol
製品番号(カタログ番号) A18389.18
または、製品番号A18389-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS543-20-4
IUPAC Namebutanedioyl dichloride
Molecular FormulaC4H4Cl2O2
InChI KeyIRXBNHGNHKNOJI-UHFFFAOYSA-N
SMILESClC(=O)CCC(Cl)=O
さらに表示
Appearance (Color)Clear pale yellow to yellow to orange
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
Refractive Index1.4675-1.4735 @ 20?C
FormLiquid
Identification (FTIR)Conforms
It is employed in the synthesis of biseugenyl succinate and bis(4-maleimidephenyl) succinate. It is also used as used as an important raw material and intermediate in pharmaceuticals, organic synthesis, agrochemicals and dye stuffs. It is also applied as hydrolytic reagent for the determination of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of biseugenyl succinate and bis(4-maleimidephenyl) succinate. It is also used as used as an important raw material and intermediate in pharmaceuticals, organic synthesis, agrochemicals and dye stuffs. It is also applied as hydrolytic reagent for the determination of water.

Solubility
Reacts violently with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, alcohols and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Johannes Karl Fink.Reactive Polymers Fundamentals and Applications: A Concise Guide to Industrial Polymers,2; William Andrew; UK, 2005; 274.
  2. R. Belcher.; J.H. Thompson.; T.S. West. Succinyl chloride as a hydrolytic reagent for the determination of water.Anal. Chim. Acta.1958,19148-153.