Benzyl nicotinate, 98%
Benzyl nicotinate, 98%
Benzyl nicotinate, 98%
Thermo Scientific Chemicals

Benzyl nicotinate, 98%

CAS: 94-44-0 | C13H11NO2 | 213.236 g/mol
製品番号(カタログ番号) A18505.18
または、製品番号A18505-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS94-44-0
IUPAC Namebenzyl pyridine-3-carboxylate
Molecular FormulaC13H11NO2
InChI KeyKVYGGMBOZFWZBQ-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)C1=CC=CN=C1
さらに表示
Identification (FTIR)Conforms
Refractive Index1.5680-1.5720 @ 20°C
FormLiquid or solidified mass
Appearance (Color)Clear yellow to very dark yellow or brown-yellow to very dark brown-yellow or orange to brown to very dark brown
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only