7-Hydroxyflavone, 98%
7-Hydroxyflavone, 98%
7-Hydroxyflavone, 98%
Thermo Scientific Chemicals

7-Hydroxyflavone, 98%

CAS: 6665-86-7 | C15H10O3 | 238.242 g/mol
製品番号(カタログ番号) A18538.06
または、製品番号A18538-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6665-86-7
IUPAC Name7-hydroxy-2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O3
InChI KeyMQGPSCMMNJKMHQ-UHFFFAOYSA-N
SMILESOC1=CC=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
さらに表示
Assay (HPLC)≥97.5%
Appearance (Color)White to cream to pale brown
Melting Point (clear melt)243.0-249.0°C
FormPowder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only