beta-Naphthoflavone, 98+%
beta-Naphthoflavone, 98+%
beta-Naphthoflavone, 98+%
Thermo Scientific Chemicals

beta-Naphthoflavone, 98+%

CAS: 6051-87-2 | C19H12O2 | 272.303 g/mol
製品番号(カタログ番号) A18543.03
または、製品番号A18543-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6051-87-2
IUPAC Name3-phenyl-1H-benzo[f]chromen-1-one
Molecular FormulaC19H12O2
InChI KeyOUGIDAPQYNCXRA-UHFFFAOYSA-N
SMILESO=C1C=C(OC2=C1C1=CC=CC=C1C=C2)C1=CC=CC=C1
さらに表示
Appearance (Color)White to cream
Assay (HPLC)≥98.0%
Melting Point (clear melt)159.0-165.0?C
FormCrystals or powder or crystalline powder
Inhibitor of aryl hydrocarbon hydroxylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only