Phenyl trans-beta-styryl sulfone, 96%
Phenyl trans-beta-styryl sulfone, 96%
Phenyl trans-beta-styryl sulfone, 96%
Phenyl trans-beta-styryl sulfone, 96%
Thermo Scientific Chemicals

Phenyl trans-beta-styryl sulfone, 96%

CAS: 16212-06-9 | C14H12O2S | 244.308 g/mol
製品番号(カタログ番号) A18556.03
または、製品番号A18556-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16212-06-9
IUPAC Name[(1Z)-2-(benzenesulfonyl)ethenyl]benzene
Molecular FormulaC14H12O2S
InChI KeyDNMCCXFLTURVLK-QXMHVHEDSA-N
SMILESO=S(=O)(C=C/C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or granules
Identification (FTIR)Conforms
Melting Point (clear melt)73-76?C
Assay (GC)≥95.0%
Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only