2-Coumaranone, 97%
2-Coumaranone, 97%
2-Coumaranone, 97%
Thermo Scientific Chemicals

2-Coumaranone, 97%

CAS: 553-86-6 | C8H6O2 | 134.134 g/mol
製品番号(カタログ番号) A18617.14
または、製品番号A18617-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS553-86-6
IUPAC Name2,3-dihydro-1-benzofuran-2-one
Molecular FormulaC8H6O2
InChI KeyACZGCWSMSTYWDQ-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2O1
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)45.0-52.0°C
Appearance (Color)White to pale cream to cream to yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only