2,4,5,6-Tetraaminopyrimidine sulfate, 97%
2,4,5,6-Tetraaminopyrimidine sulfate, 97%
2,4,5,6-Tetraaminopyrimidine sulfate, 97%
Thermo Scientific Chemicals

2,4,5,6-Tetraaminopyrimidine sulfate, 97%

CAS: 5392-28-9 | C4H10N6O4S | 238.22 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A18788.09
または、製品番号A18788-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5392-28-9
IUPAC Namepyrimidine-2,4,5,6-tetramine; sulfuric acid
Molecular FormulaC4H10N6O4S
InChI KeyMQEFDQWUCTUJCP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
さらに表示
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0% (Dry wt. basis)
Loss on Drying≤9% (1g, 150°C, 3hrs)
Appearance (Color)Ochre to brown
FormCrystalline powder or powder
Used as an intermediate in the production of Methotrexate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an intermediate in the production of Methotrexate.

Solubility
Soluble in water and dimethyl sulfoxide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Chiosis G; Lucas B; Shtil A; Huezo H; Rosen N. Development of a purine-scaffold novel class of Hsp90 binders that inhibit the proliferation of cancer cells and induce the degradation of Her2 tyrosine kinase. Bioorg Med Chem., 2002, 10 (11), 3555-64.
  2. Palanki, M., et al. Discovery of 3,3‘-(2,4-Diaminopteridine-6,7-diyl)diphenol as an Isozyme-Selective Inhibitor of PI3K for the Treatment of Ischemia Reperfusion Injury Associated with Myocardial Infarction. J. Med. Chem., 2007, 50(18), 4279-4294.