2,4,6-Trichloroaniline, 98+%
2,4,6-Trichloroaniline, 98+%
2,4,6-Trichloroaniline, 98+%
Thermo Scientific Chemicals

2,4,6-Trichloroaniline, 98+%

CAS: 634-93-5 | C6H4Cl3N | 196.455 g/mol
製品番号(カタログ番号) A19044.14
または、製品番号A19044-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS634-93-5
IUPAC Name2,4,6-trichloroaniline
Molecular FormulaC6H4Cl3N
InChI KeyNATVSFWWYVJTAZ-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=C(Cl)C=C1Cl
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)74.0-81.0?C
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. B. T. Gowda,; I. Svoboda ,; H. Fuess. 2,4,6-Trichloro­aniline. Acta Cryst. 2007 , 63o3347.
  2. Mihály Kádára,; Zoltán Nagy,; Tamás Karancsi,; György Farsang. The electrochemical oxidation of 4-chloroaniline, 2,4-dichloroaniline and 2,4,6-trichloroaniline in acetonitrile solution. Electrochimica Acta. 1991, 46 (9),1297-1306.