Thermo Scientific Chemicals

10-Undecenoic acid, 99%, Thermo Scientific Chemicals

產品號碼: A19122.0B
1000 g, Each
Thermo Scientific Chemicals

10-Undecenoic acid, 99%, Thermo Scientific Chemicals

產品號碼: A19122.0B
1000 g, Each
Quantity
產品號碼: A19122.0B
亦稱為 A19122-0B
價格 (TWD)

化學識別

CAS
112-38-9
IUPAC Name
undec-10-enoic acid
Molecular Formula
C11H20O2
InChI Key
FRPZMMHWLSIFAZ-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCCC=C
Appearance (Color)
Colorless to white to pale yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Fused solid or clear liquid as melt
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms

說明

10-Undecenoic acid is used for the preparation of active pharmaceutical ingredients, cosmetics, perfumes, antidandruff shampoos and antimicrobial powders. It is used for the treatment of skin problems and antifungal agent. It is involved in the production of nylon-11, methyl undecylenate, undecylenic aldehyde and undecylenic alcohol. Further, it is used as musk in perfumes and aromas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecenoic acid is used for the preparation of active pharmaceutical ingredients, cosmetics, perfumes, antidandruff shampoos and antimicrobial powders. It is used for the treatment of skin problems and antifungal agent. It is involved in the production of nylon-11, methyl undecylenate, undecylenic aldehyde and undecylenic alcohol. Further, it is used as musk in perfumes and aromas.

Solubility
Soluble in alcohol, chloroform, diethyl ether, benzene and fixed or volatile oils. Insoluble in hot water and cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text