Ethyl 3-benzoylacrylate, predominantly trans, 94%
Ethyl 3-benzoylacrylate, predominantly trans, 94%
Ethyl 3-benzoylacrylate, predominantly trans, 94%
Thermo Scientific Chemicals

Ethyl 3-benzoylacrylate, predominantly trans, 94%

CAS: 17450-56-5 | C12H12O3 | 204.225 g/mol
製品番号(カタログ番号) A19426.09
または、製品番号A19426-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS17450-56-5
IUPAC Nameethyl (2E)-4-oxo-4-phenylbut-2-enoate
Molecular FormulaC12H12O3
InChI KeyACXLBHHUHSJENU-CMDGGOBGSA-N
SMILESCCOC(=O)C=CC(=O)C1=CC=CC=C1
さらに表示
Refractive Index1.5415-1.5485 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥92.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only