4-(2-Furyl)-3-buten-2-one, cis + trans, 98%
4-(2-Furyl)-3-buten-2-one, cis + trans, 98%
4-(2-Furyl)-3-buten-2-one, cis + trans, 98%
Thermo Scientific Chemicals

4-(2-Furyl)-3-buten-2-one, cis + trans, 98%

CAS: 623-15-4 | C8H8O2 | 136.15 g/mol
製品番号(カタログ番号) A19456.14
または、製品番号A19456-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS623-15-4
IUPAC Name(3E)-4-(furan-2-yl)but-3-en-2-one
Molecular FormulaC8H8O2
InChI KeyGBKGJMYPQZODMI-SNAWJCMRSA-N
SMILESCC(=O)\C=C\C1=CC=CO1
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)Pale yellow to orange to brown
Melting Point (clear melt)35.0-41.0°C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only