4-(Trifluoromethoxy)phenylacetic acid, 98%
4-(Trifluoromethoxy)phenylacetic acid, 98%
4-(Trifluoromethoxy)phenylacetic acid, 98%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenylacetic acid, 98%

CAS: 4315-07-5 | C9H6F3O3 | 219.14 g/mol
製品番号(カタログ番号) A19512.03
または、製品番号A19512-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4315-07-5
IUPAC Name2-[4-(trifluoromethoxy)phenyl]acetate
Molecular FormulaC9H6F3O3
InChI KeyZMTIBXQPXBUWPQ-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC=C(OC(F)(F)F)C=C1
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)83.0-89.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only