Ethyl thiophene-3-acetate, 98%
Ethyl thiophene-3-acetate, 98%
Ethyl thiophene-3-acetate, 98%
Ethyl thiophene-3-acetate, 98%
Thermo Scientific Chemicals

Ethyl thiophene-3-acetate, 98%

CAS: 37784-63-7 | C8H10O2S | 170.23 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A19694.22
または、製品番号A19694-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS37784-63-7
IUPAC Nameethyl 2-(thiophen-3-yl)acetate
Molecular FormulaC8H10O2S
InChI KeyFZBNQIWPYCUPAP-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CSC=C1
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.5080-1.5120 @ 20?C
Assay (GC)≥97.5%
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kaewtong, C.; Niamsa, N.; Wanno, B.; Morakot, N.; Pulpoka, B.; Tuntulani, T. Optical chemosensors for Hg 2+ from terthiophene appended rhodamine derivatives: FRET based molecular and in situ hybrid gold nanoparticle sensors. New J. Chem. 2014, 38 (8), 3831-3839.
  2. Pardieu, E.; Saad, A.; Dallery, L.; Garnier, F.; Vedrine, C.; Hauquier, F.; Dalko, P.; Pernelle, C. Synthesis and characterization of β-substituted 3, 4-ethylenedioxy terthiophene monomers for conducting polymer applications. Synt. Met. 2013, 171, 23-31.