Tridecanal, 96%, stab.
Tridecanal, 96%, stab.
Tridecanal, 96%, stab.
Thermo Scientific Chemicals

Tridecanal, 96%, stab.

CAS: 10486-19-8 | C13H26O | 198.35 g/mol
製品番号(カタログ番号) A19722.06
または、製品番号A19722-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10486-19-8
IUPAC Nametridecanal
Molecular FormulaC13H26O
InChI KeyBGEHHAVMRVXCGR-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC=O
さらに表示
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
Identification (FTIR)Conforms
FormLiquid
Free acid (titration)≤1.5%
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only