Methyl 5-methylisoxazole-3-carboxylate, 97%
Methyl 5-methylisoxazole-3-carboxylate, 97%
Methyl 5-methylisoxazole-3-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 5-methylisoxazole-3-carboxylate, 97%

CAS: 19788-35-3 | C6H7NO3 | 141.13 g/mol
製品番号(カタログ番号) A19814.14
または、製品番号A19814-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19788-35-3
IUPAC Namemethyl 5-methyl-1,2-oxazole-3-carboxylate
Molecular FormulaC6H7NO3
InChI KeyMVHHQOCEOUNTID-UHFFFAOYSA-N
SMILESCOC(=O)C1=NOC(C)=C1
さらに表示
Melting Point (clear melt)87.0-96.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only