Thermo Scientific Chemicals

Sodium sulfate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: A19890.0I
5000 g, Each
Thermo Scientific Chemicals

Sodium sulfate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: A19890.0I
5000 g, Each
Quantity
Catalog number: A19890.0I
also known as A19890-0I
Price (USD)
Price: 169.00
Online price: 143.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7757-82-6
IUPAC Name
disodium sulfate
Molecular Formula
Na2O4S
InChI Key
PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S([O-])(=O)=O
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥98.5%
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≤0.5%

Description

Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.

Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text