Morpholine-4-carboxamidine hydrobromide, 98%
Morpholine-4-carboxamidine hydrobromide, 98%
Morpholine-4-carboxamidine hydrobromide, 98%
Thermo Scientific Chemicals

Morpholine-4-carboxamidine hydrobromide, 98%

CAS: 157415-17-3 | C5H12BrN3O | 210.075 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A19985.22
または、製品番号A19985-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS157415-17-3
IUPAC Namemorpholine-4-carboximidamide hydrobromide
Molecular FormulaC5H12BrN3O
InChI KeyIOHSDHMCZXCESZ-UHFFFAOYSA-N
SMILESBr.NC(=N)N1CCOCC1
さらに表示
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Xiaohu Deng, et al. CuI-Catalyzed Amination of Arylhalides with Guanidines or Amidines: A Facile Synthesis of 1-H-2-Substituted Benzimidazoles.J. Org. Chem.,2009,74(15), 5742-5745.