GeneArt™ Genomic Cleavage Selection Kit - FAQs
GeneArt™ Genomic Cleavage Selection Kit - FAQs
View additional product information for GeneArt™ Genomic Cleavage Selection Kit - FAQs (A27663)
68 product FAQs found
What are TALs or TALENS?
TALs or TALENs are transcription activator-like effector nuclease proteins that are naturally occurring transcriptional activators secreted by Xanthomonas spp. into their plant hosts. GeneArt TALs are derived from Xathomonas TAL effectors, the DNA-binding domain of which consists of a variable number of amino acid repeats. Each repeat contains 33–35 amino acids and recognizes a single DNA base pair. The DNA recognition occurs via 2 hypervariable amino acid residues at positions 12 and 13 within each repeat, called repeat-variable di-residues (RVDs). TAL effector repeats can be assembled in modular fashion, varying the RVDs to create a TAL protein that recognizes a specific target DNA sequence.
I'm hoping to use one set of TALs to knockout two genes. I have identified potential binding sites, but I could not find a spot with 100% homology between the two genes. Will the TAL work if there is a 1 or 2 bp mismatch in the binding domain?
We have observed that as little as 3 bp mismatch affects the binding of the TALs. We recommend that there be an exact match between the design and the binding site. If you would like some help with the design, please email geneartsupport@lifetech.com.
I just received my Invitrogen GeneArt TALs vectors. Can I store them lyophilized?
We recommend that you resuspend the vector in 50 µL of distilled water or 10 mM Tris-HCl (pH 8.0) and incubate for 1 hour at room temperature. Resuspend the vector DNA by gently pipetting up and down 5-10 times. Store the resuspended DNA at -20°C.
I have questions regarding licensing of TALs that is not covered under the Limited-Use Label License (LULL). Whom should I contact?
Please email our team, at outlicensing@lifetech.com.
Do I need to use a specific transfection reagent when working with a TAL effector vector?
No, we would recommend either lipid-mediated transfection or electroporation. Please review relevant references or consult with the supplier of your cell line for the optimal method of transfection.
How can an Invitrogen GeneArt Precision TAL protein fused to a Fok1 endonuclease be used to induce gene silencing or to insert an engineered DNA fragment into an exact location in the genome?
Double-stranded DNA breaks can be created at your specified genomic locus by using a pair of Invitrogen GeneArt Precision TAL proteins that have been fused to the Fok1 endonuclease. Using a pair of Precision TAL proteins for the targeting reduces off-target effects. The breaks induced by the Fok1 nuclease domain are subsequently repaired through either of two endogenous cellular mechanisms: nonhomologous end joining (NHEJ), or homology-directed repair (HDR). NHEJ is prone to errors and often introduces a frameshift mutation when it occurs within the coding sequence of a protein-coding gene, effectively silencing the gene. Homologous DNA “donor sequences” can be used with HDR to introduce a defined new DNA sequence. Consequently, a Precision TAL protein fused to a Fok1 endonuclease can be used to induce gene silencing or to accurately insert an engineered DNA fragment into an exact location in the genome.
When should I use a native DNA-binding domain compared to a truncated one for TALs?
Truncated TAL Fok1: Recommended for mammalian cells
Native TAL Fok1: Recommended for plants
Native Activators: Higher performance in non-mammalian systems
Truncated MCS: Removing endogenous activator activity
How are TALs expressed?
All TALs are expressed as Entry clones (Gateway vector-compatible, flanked by attL1 and attL2).
Fok1 TALs:
- TALs tethered with the Fok1 nuclease may be used for targeting specific genes for silencing.
- Fok1 is a Type IIs restriction endonuclease from Flavobacterium okeanokoites, consisting of an N-terminal DNA-binding domain and a C-terminal nonspecific DNA cleavage domain.
- Fok1 acts as a nuclease pair and binds to the DNA duplex at target sites designated by the binding domains resulting in subsequent cleavage.
VP16 or VP64 TALs:
- Either VP16 or VP64 may be used to increase endogenous or recombinant gene expression levels.
- VP16 is a trans-acting protein originating from the herpes simplex virus, to form a complex with host transcription factors to induce immediate early gene transcription.
- VP64 is a tetrameric form of the VP16 minimal activation domain.
MCS TALs:
- The multiple cloning site (MCS) TAL allows the user to clone any desired effector domain for targeting to any locus within the genome.
How do I transfect TALs?
Please follow standard plasmid DNA recommendations for transfection. We recommend our Invitrogen Lipofectamine 3000 and Invitrogen Lipofectamine 2000 reagents.
What is the TAL workflow?
- Design Invitrogen GeneArt TALs for target DNA, and clone into Gateway entry or destination vectors
- In vitro validation of TALs (optional)
- Transfect cells with TAL expression vectors
- TAL-mediated target cleavage
- Cleavage analysis using Invitrogen GeneArt Genomic Cleavage Detection Kit
Are TAL-meditated KO or KI strains considered GMO (genetically modified organisms)?
Yes, KO and KI strains involve editing the native genetic code by either mutating or deleting an encoded message or inserting a new piece of information at a desired site. Although this does manipulate the native genetic information, this technology, when used in a responsible manner, has very useful applications, including engineering yeasts for insulin production or engineering cells for more economically and clinically valuable products.
What is the best delivery method for TALs? Transfection, electroporation, or retroviral delivery?
mRNA and DNA are best delivered via lipid-based transfection for standard test cell lines (i.e., 293, HeLa, etc.). mRNA delivery also reduces the risk of transgene integration. We offer products including our Invitrogen Lipofectamine MessengerMAX Reagent for delivery of mRNA, and Invitrogen Lipofectamine 3000 Reagent for delivery of DNA. For stem cells, electroporation is the best option.
How fast can I get TAL constructs made so that I can do my KO or KI experiments?
Manufacturing takes place typically within 2 weeks after your order has been received.
Can TALs recognize degenerate binding sites?
By careful designing they can be engineered to be very specific. Recent publications show that 1-3 bp mismatches in target DNA sequences can be tolerated to a large extent.
Can TALs be delivered by retroviruses?
Yes. If viral-based delivery is your preferred option, we recommend adenoviral systems over lentiviral systems for TAL delivery.
How big is the TAL vector construct?
The TAL vector construct is 3.3 kb.
How specific is TAL/TALEN technology? How about off-site targeting?
A recent paper (http://www.nature.com/nbt/journal/v31/n9/abs/nbt.2675.html) by Prashant Mali in Nature Biotechnology shows that TALENs are tolerant of 1-2 mismatches, but less tolerant to a large majority of 3 bp mismatches.
What vectors are offered with the Invitrogen GeneArt PerfectMatch TALS?
We offer the N-TAL Fok1 Entry Gateway vector or the Fok1 CMV promoter–driven vector for expression.
What is the difference between Invitrogen GeneArt Precision TALs and Invitrogen GeneArt PerfectMatch TALs?
Invitrogen GeneArt PerfectMatch TALs are derived from Invitrogen GeneArt Precision TALs. PerfectMatch TALs have a new design that removes the 5′ base constraint, and therefore, can be designed to target any desired sequence in the genome. They contain 3 amino acids mutated at the N terminus of the TAL effector, which converts the 5′ binding motif to a universal binding motif able to bind to any base: A, G, C, or T. These PerfectMatch TALs can be designed with Fok1 nuclease in a Invitrogen Gateway entry vector or with CMV-driven expression for ready-to-express format for mammalian systems. PerfectMatch TALS perform as well as or better than our original Precision TALs. Currently, PerfectMatch TALs are only available with nuclease function (Fok1 Nuclease Pair), whereas Precision TALs are offered with nuclease function (Fok1 Nuclease Pair), activator function (VP16 or VP64), or custom function (MCS vector).
We compared cleavage efficiencies of PerfectMatch and Precision TALs designed for the HPRT locus using the Invitrogen GeneArt Genomic Cleavage Detection Kit, and found PerfectMatch TALs exhibit cleavage efficiencies equal to or better than the performance of Precision TALs on the same targeted region.
PerfectMatch TALs increase the flexibility of designing TAL effector targets and make it possible to keep the spacing distance between targets of TAL effector pairs at 15–16 bp to get maximal TAL effector efficiency.
What guidelines should I follow when designing a TAL sequence?
Here are our suggestions:
Invitrogen GeneArtPrecision TALs allow the construction of TAL effector functional proteins directed to either 18- or 24-base DNA target sites.
Each target site must be preceded by a 5′ T because the N terminus of the TAL effector protein contains a conserved T-binding motif. The 5′ T does not count as one of the 18 or 24 bases to be selected for targeting your specific site.
Nuclease pairs need to be designed with a spacing of 13-18 bp between the target sites on opposite strands of the DNA.
For Invitrogen GeneArt PerfectMatch TALs, there are no restrictions for the 5′ base. We developed these second-generation TALs by mutating the N-terminal domain to reduce its specificity for 5′ T. Therefore, any 5′ base (T, G, C, or A) can be used with performance comparable to that of the original Precision TALs. We recommend that you design nuclease pairs with a 15-16 bp spacing between the two TAL effectors.
The contribution of individual binding motifs within the DNA-binding domain to TAL effector binding efficiency is thought to differ, since strong and weak binding motifs exist. The A- and T-binding motifs are thought to fall within the “weak binder” category, while the C- and G-binding motifs are thought to be “strong binders”. Stretches of more than 5 weak binders should be avoided at the extreme 5′ end of the binding domain (not counting the 5′ T), or if they are not flanked by Cs. We recommended that you select a TAL effector with a DNA-binding domain composed of mixed binding motifs for best results. In the context of the living cell, DNA accessibility also determines TAL effector efficiency. It is possible that chromatin, DNA methylation, and/or proteins bound to the DNA may interfere with TAL binding. Although promoter structure varies, and specific rules regarding design are currently lacking, it is recommended that TAL transcription factors used for transcriptional activation of natural promoters be positioned upstream of the TATA box, or in some cases downstream of the transcriptional start site. Selecting a target site directly over the TATA box or other known transcription factor binding site is not recommended. Be sure that the natural ATG is present, and that no premature ATG which may interfere with the natural translational start is transcribed.
What available functionality (effector domains) do you offer for TALs?
Please view the available effector domains (https://www.thermofisher.com/us/en/home/life-science/cloning/gene-synthesis/geneart-precision-tals.html#table1).
What is the turnaround time for receiving custom TALs?
We will ship you a clone with a verified, optimized sequence approximately 2 weeks after confirming your order.
What are the benefits of using TAL/TALEN-based genome editing compared to your CRISPR system?
Invitrogen GeneArt Precision TALs, in addition to gene deletion, down-regulation and integration, can also be used for gene activation. Additionally, the system is based on a protein-DNA system, in contrast to CRISPR, which is based on a RNA-DNA system. TALs can be used to target any gene in any cell, including mammalian, bacterial, yeast, plants, insect, stem cells and zebrafish. Lastly, off-target effects are low when using the TAL system. Please refer to the following paper (http://www.sciencedirect.com/science/article/pii/S016816561500200X) where the authors compared TALs and CRISPR technology.
Why should I use TAL/TALEN technology?
- One-to-one relationship between two critical amino acids in each repeat and each DNA base in the target sequence
- Simple code for creating engineered TALs
- More predictable than zinc fingers
- Modular assembly of domains allows engineering of sequence-specific DNA-binding proteins
- Can be coded to deliver functionality to a specific locus for: nucleases, activators, repressors, chromatin modifiers
My results do not match data from the GeneArt Genomic cleavage selection assay. Why is this?
Results are locus-dependent. We recommend using the Invitrogen GeneArtGenomic Cleavage Detection Kit (Cat. No. A24372) to verify cleavage on the endogenous genomic locus.
I am using the GeneArt Genomic Cleavage Selection Kit and am seeing too much OFP background. What can I do?
Too much OFP background can occur if there is plasmid contamination, and/or can be cell line- and target-dependent. Make sure to pick single clones when culturing the cleavage selection plasmid, or try to reduce the amount of vector that is included in the transfection.
When using the GeneArt Genomic Cleavage Selection Kit, no OFP is observed. What should I do?
This can occur if:
- Oligos are designed incorrectly; make sure to include specific sequences for TALEN/CRISPR target.
- Transfection efficiency is too low; optimize conditions or use Invitrogen Lipofectamine 3000 or 2000 reagent for best results.
- Cell line-dependent; OFP does not express well in certain cell lines; we recommend using 293FT cells as a test for cleavage activity.
I am using the GeneArt Genomic Cleavage Selection Kit and am getting very few ampicillin-resistant colonies on the selection plate. What should I do?
Here are possible reasons and solutions:
- Single-stranded (ss) oligonucleotides designed incorrectly; Make sure that each ss oligonucleotide contains the 4 nucleotides on the 5′ end required for cloning into the Invitrogen GeneArt Genomic Cleavage Selection vector: ie., Top strand includes AATT on the 5′ end; Bottom strand includes CTAG on the 5′ end.
- Double-stranded (ds) oligonucleotides were degraded; Store the 50 nM ds oligonucleotide stock in 1X ligation buffer at -20°C. Avoid repeat freeze/thaw cycles, aliquot 50 nM ds oligonucleotides stock and store at -20°C.
- Oligonucleotide annealing reaction was inefficient: Ensure that the annealing reaction was performed as directed. If ambient temperature is >25°C, incubate the annealing reaction in a 25°C incubator.
How can I increase my efficiency of genome editing using the CRISPR-Cas9 technology?
There are several ways to increase efficiency, for instance, adding antibiotic selection and/or FAC sorting to enrich for the transfected cells will both help.
I am interested in using the CRISPR-Cas9 technology for genome editing. However, there is no 5’ NGG (PAM) sequence close to my locus of interest. What can I do?
PAM is a necessary requirement for CRISPR gene editing. However, in its absence, we recommend engineering a TAL effector to edit your desired gene efficiently. We offer GeneArt PerfectMatch TAL effectors. These are TAL effector nucleases that remove the 5´ base constraint and can be designed to target any desired sequence within the genome. Please go here for further details: https://www.thermofisher.com/us/en/home/life-science/genome-editing/geneart-tals.html
What is the peak excitation and emission of OFP? What type of laser should I use?
OFP has peak excitation of 548 nm and emission of 560 nm. A 488 nm laser is recommended for efficient excitation. Standard 530/30, 574/26 and 603/48 emission filters are recommended for detection.
Can the GeneArt Genomic Cleavage Selection Kit be used to verify cleavage of the endogenous genomic locus?
OFP and CD4 expression are considered an estimation rather than absolute quantification of genomic cleavage. We recommend using our GeneArt Genomic Cleavage Detection Kit to verify cleavage on endogenous genomic locus
How can I enrich for the nuclease-modified cells using the GeneArt Genomic Cleavage Selection Kit?
The Invitrogen GeneArt genomic cleavage selection vector also contains the membrane protein CD4 coding gene that is fused with OFP through the T2A self-cleavage peptide, allowing nuclease-modified cells to be enriched through cell sorting or CD4 antibody-conjugated Dynabeads. This cleavage selection vector allows simple, rapid evaluation of the functionality of the programmable nuclease, and direct enrichment of the genome-modified cells.
How soon can you detect cleavage by TAL or CRISPR-Cas9 systems using the GeneArt Genomic Cleavage Selection Kit?
It is possible to verify cleavage as early as 24 hours post-transfection by checking for OFP expression of the transfected cells under the microscope. The percentage of OFP-positive cells indicates the cleavage activity of TAL or CRISPR-Cas9.
What is the difference between the GeneArt Genomic Cleavage Selection Kit and the GeneArt Genomic Cleavage Detection Kit?
GeneArtGenomic Cleavage Selection Kit
- Fast, live detection
- Visual indication (fluorescence)
- Proves editing tool works
- Allows for clone enrichment
GeneArt Genomic Cleavage Detection Kit
- Requires cells to be lysed
- Quantifiable results
- Negative result does not indicate whether editing tool works or not
- No enrichment capabilities
Can the GeneArt Genomic Cleavage Selection Kit be used to evaluate TALs and/or CRISPR constructs?
Yes, the GeneArt Genomic Cleavage Selection Kit can be used with either TALs or CRISPR constructs.
How does the GeneArt Genomic Cleavage Selection Kit work?
The vector in the kit has an OFP gene that is interrupted by the insertion of a cloning site for the target sequence of the programmable nuclease. The upstream sequence coding for the N-terminal portion of the OFP gene contains a region complementary to the 5′ end of the C-terminal region of the OFP gene. A stop codon after the N-terminal OFP sequence ensures no expression of the reporter prior to nuclease activity. The CD4 gene is out of frame for expression when the OFP gene is interrupted by the cloning site. Double-stranded breaks cause the complementary strands of the end sequences of the OFP gene to recombine, and OFP expression is restored. The CD4 gene is now in frame for expression and can be screened for via FACs analysis or Dynabeads magnetic bead selection.
What is the GeneArt Genomic Cleavage Selection Kit?
The GeneArt Genomic Cleavage Selection Kit can be used to:
- Screen for functionality of your engineered nucleases as early as 24 hours post-transfection using fluorescence microscopy
- Enrich for modified cells using fluorescence-activated cell sorting (FACS) or Dynabeads CD4 Magnetic Beads
Can I perform multiplexed transfection of CRISPR gRNAs using the Invitrogen Neon Transfection System?
Yes, the Neon system does work for multiple gRNAs transfected at the same time.
What is the suggested ratio of Cas9 mRNA to IVT gRNA?
We recommend starting at a ratio of 0.5 µg of Cas9 mRNA and 50 ng of each IVT gRNA per well in a 24-well format. You should determine the optimal ratio for your particular cell line via a dose-response study.
I am interested in multiplexing. How can I do this using the GeneArt CRISPR system?
Create multiple gRNAs targeting the targets of your choice, followed by co-transfection with GeneArt CRISPR Nuclease mRNA or GeneArt Platinum Cas9 Nuclease. To make the gRNAs for Cas9 mRNA, use GeneArt CRISPR Strings DNA, U6 or IVT gRNAs (generated using either GeneArt CRISPR Strings DNA, T7 or the GeneArt Precision gRNA Synthesis Kit). For the Cas9 protein, use IVT gRNAs (generated using either GeneArt CRISPR Strings DNA, T7 or the GeneArt Precision gRNA Synthesis Kit).
Is it possible to insert a selection marker, e.g., neomycin or a fluorescent marker, into a preexisting CRISPR-Cas9 plasmid?
Yes, if you use the current Invitrogen GeneArt CRISPR nuclease vectors the respective Limited-Use Label Licenses (LULLs) will apply.
I see other publications using different PAM sequences such as NNNGATT, NNAGAA, or NAAAAC. Can I use that instead of NGG?
No, the PAM sequence is unique to the bacterial species that was used to create the Cas9. In the Invitrogen GeneArt kits, we derived Cas9 from Streptococcus pyogenes.
I am using the CRISPR-Cas9 technology. What is the PAM sequence?
PAM stands for the protospacer adjacent motif. It is necessary for Cas9 to bind to the DNA successfully. The PAM sequence for the Streptococcus pyogenes Cas9 in the Invitrogen GeneArt CRISPR kits is NGG.
I am interested in using the CRISPR system to modify my gene of interest using HDR instead of NHEJ repair. How can I do this?
With the CRISPR-Cas9 editing complex (DNA vector, mRNA or Protein), co-transfect a DNA repair template that contains high homology to the sequence of interest along with the desired sequence you would like to introduce into the DNA. By doing so HDR can occur, and your specific edits (mutation, insertion, etc.) can be incorporated into the genome.
How can I tell if my CRISPR genome editing experiment is working?
Cleavage efficiency can be detected using the Invitrogen GeneArt Genomic Cleavage Detection Assay. This assay relies on mismatch detection endonucleases to detect insertions and deletions (indels) generated during cellular NHEJ repair.
What is an indel?
An indel refers to the genomic insertion or deletion of bases, which are incorporated during either cellular NHEJ or HD repair mechanisms.
Can you elaborate on your suggestion to use CRISPR as a screening tool before going to TALs?
Since cleavage efficiency at a particular locus depends on the accessibility of the locus, chromatin state, and sequence, it is advisable to test multiple different loci/regions within a gene of interest. With CRISPR-Cas9-mediated genome editing, for each target of interest the user needs only to change the 19-20 bp target-specific oligo. After the cell lines have been screened and the sequence/locus with the highest cleavage efficiency has been identified, the biologically relevant mutations can be precisely created with high-specificity Invitrogen GeneArt TALs (https://www.thermofisher.com/us/en/home/life-science/genome-editing/geneart-tals.html).
I am using the CRISPR-Cas9 technology. How precise is the cleavage event?
Cleavage is precise, and, after binding of the Cas9 and gRNA complex to the target genomic sequence, the nuclease activity occurs 3 bases upstream of the PAM (NGG) site.
I am using the CRISPR technology to knock out a gene. How long after transfection should I assess mRNA levels and protein knockdown?
This would depend upon the half-life of the particular transcript in your cell. We typically start seeing reduction in mRNA levels as early as 24 hrs post transfection, with further reduction after 48-72 hrs. Hence, we recommend performing the genomic cleavage detection assay 48-72 hours post transfection.
What happens to Cas9 after it performs its endonuclease activity?
Cas9 is transiently expressed and will therefore disappear over time with successive cell divisions.
Is it possible to use CRISPR-Cas technology for prokaryotic gene engineering?
Yes, it is possible but our system is not for prokaryotes, and has only been optimized for mammalian systems. Please also consult our CRISPR custom services for further inquiries (custom.services@lifetech.com).
Have you tested the CRISPR system in hosts other than mammalian systems?
We have only tested these in mammalian systems (human and mouse cells).
Do you offer a service to generate custom cell lines using CRISPR?
Yes, we do offer this service (https://www.thermofisher.com/us/en/home/life-science/genome-editing/genome-engineering-services/cell-line-engineering-services.html).p>
I am using the CRISPR technology. I would like to design a guide RNA (gRNA) to insert a selection cassette such as neomycin. How can I do this?
The gRNA oligo design strategy in the Invitrogen GeneArt CRISPR Nuclease User Guide (https://tools.thermofisher.com/content/sfs/manuals/GeneArt_CRISPR_nuclease_mRNA_man.pdf)describes how you can design the guide RNA to target the locus in which the neomycin cassette should be inserted. The cassette (neomycin) can be inserted via HDR, in which case the neomycin cassette should contain locus specific homology arms.
I am trying to target a big and multifunctional protein using CRISPR technology. To what site should I design my sgRNA? How can I check that the editing occurred?
The first few exons would be best (closer to the promoter, resulting in premature transcript termination). Since the gRNA efficiency depends on the accessibility of the locus as well as the chromatin structure at that location, it is advisable to design and test a few target sites. Non-CRISPR-related mutations may be identified using gDNA isolated from non-CRISPR-treated cells as a control and performing a Invitrogen GeneArt Genomic Cleavage Detection Assay (https://www.thermofisher.com/order/catalog/product/A24372). Standard western blot analysis is a good measure for the verification of protein levels.
Can I use CRISPR technology to introduce large sequences, such as GFP or IRES?
Yes, this should be possible using CRISPR technology combined with HDR.
I am using the CRISPR technology and am worried about off-target effects. What is the best way to combat this issue?
Carefully designed crRNA target oligos and avoiding homology with other regions in the genome are critical for minimizing off-target effects.
What is the efficiency of HDR (homology directed repair) after the double-stranded break?
HDR efficiency is very low, on average less than 2%.
Can you provide suggestions on how to introduce a fragment into the genome or important sequence by HDR (homology directed repair)?
Create a double-stranded DNA break using the GeneArt CRISPR Nuclease Vector (https://www.thermofisher.com/us/en/home/life-science/genome-editing/geneart-crispr/crispr-nuclease-vector.html), while simultaneously transfecting your plasmid-based donor repair template. Your donor repair template plasmid will contain the sequence you wish to introduce that is flanked by at least 500 bp (or more) of sequence, which results in efficient homologous recombination of your sequence.
What is most efficient for HDR (homology directed repair), and what length is recommended for the homologous exogenous DNA, single-stranded oligos, double-stranded oligos, or large homologous arms (>1 kb) from a plasmid?
All of them may work, but for better efficiency, a longer homology arm is better (at least 500 bp (or more) on either side of the exogenous DNA). The homology length is dependent on the size of the fragment and will need to be tested. ssDNA may be error-prone or choose NHEJ. We offer the Invitrogen GeneArt Strings dsDNA fragments (1-3 kb) to assist with this type of application.
What is the difference between NHEJ- and HDR-mediated repair?
Both HDR (homology directed repair) and NHEJ (non-homologous end joining) are cellular mechanisms through which double-stranded DNA lesions are repaired. When a repair template is not present, NHEJ occurs to ligate double-stranded breaks, leaving behind insertion/deletion (indel) mutations. HDR is an alternative repair pathway in which a repair template is used to copy the sequence to the double-stranded break. You can, therefore, introduce specific nucleotide changes or DNA fragments into your target gene by using HDR with a repair template.
Are the indel events that happen in a particular locus identical between the cells? Can we include a clonal step before expanding and enriching the population?
Clonal isolation and a combined cleavage analysis and sequence verification of the edited clone is advisable.
How does the Invitrogen GeneArt CRISPR-Cas system work?
As a simple two-component system that includes the Cas9 endonuclease and a noncoding guide RNA (gRNA), the engineered Type II CRISPR/Cas system can be leveraged to cleave genomic DNA at a predefined target sequence of interest. The gRNA has two molecular components: a target-complementary CRISPR RNA (crRNA) and an auxiliary trans-activating crRNA (tracrRNA). Both the gRNA and the PAM (NGG) motif guide the Cas9 nuclease to a specific genomic sequence to form a complex, followed by local strand separation (R-loop), at which the Cas9 nuclease creates a double-stranded DNA break (DSB) 3 nucleotides upstream from the PAM site. As a result, you may bring new functionality to the gene of interest via mutations, create knockouts, or introduce nonnative or synthetic genomic sequences to investigate novel applications.
CRISPR also allows for non-editing application flexibility such as gene regulation or RNAi-related studies. The Cas9 nuclease may be tethered to different functional domains (activators or repressors) or the gRNA may be designed to directly cleave miRNA.
What is the advantage of generating gene knockouts by TAL or CRISPR compared to vector-based stably expressed shRNA?
TAL and CRISPR directly edit the genome by a combined cleavage and repair mechanism to impart permanent genomic change (deletion or frameshift mutation), and the resulting gene knockouts are very efficient. RNAi technology, on the other hand, is an indirect method in either down-regulating or shutting down a gene completely through direct interaction with RNA (coding or noncoding). Even in the case for stably expressed miRNA or shRNA systems, it may be difficult to effect complete penetrance (i.e., shRNA:mRNA ratio) since knock-down levels are dependent on the activity of the promoter (related to integration location).
What is CRISPR technology used for?
With their highly flexible but specific targeting, CRISPR-Cas systems can be manipulated and redirected to become powerful tools for genome editing. CRISPR-Cas technology permits targeted gene cleavage and gene editing in a variety of eukaryotic cells, and because the endonuclease cleavage specificity in CRISPR-Cas systems is guided by RNA sequences, editing can be directed to virtually any genomic locus by engineering the guide RNA sequence and delivering it along with the Cas endonuclease to your target cell.
What does CRISPR and CRISPR-Cas stand for?
CRISPR stands for clustered regularly interspaced short palindromic repeat; CRISPR-Cas (CRISPR-associated) systems are used for genome editing in various host organisms.