Qualification Services Plan for iBright™ Imaging Systems
実際の製品は異なる場合があります
Thermo Scientific™
Qualification Services Plan for iBright™ Imaging Systems
製品番号(カタログ番号) A35425
価格(JPY)
-
Qualification services for Invitrogen iBright CL and FL imaging systems are designed to help balance business and laboratory requirements and help you comply with global standards, country-specific regulations, and other laboratory site requirements.
Verify instrument performance and minimize downtime with IQ/OQ services
Complete with audit-ready documentation, our factory-trained and experienced engineers will conduct installation qualification (IQ) and operational qualification (OQ) on the iBright imaging systems in your lab. These qualification services help ensure your instruments are installed and operating according to manufacturer’s specifications and help move you into production faster.
When is IQ/OQ recommended?
• At installation
• After a move
When is OQ recommended?
• After planned maintenance
• After major repair
• After additions, changes, or upgrades to the system
• For a scheduled OQ site requirement
For more detailed information, view the figure "Maintaining a qualified Invitrogen iBright Imaging System" below and visit instrument IQ/OQ/IPV.
Certain exclusions apply. Contact your Services and Support Representative for details at compliancesales@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.
仕様
Product LineiBright™
Product SizeOQ
Unit SizeOQ