Pierce™ 3x DYKDDDDK Peptide
Thermo Scientific™
Pierce™ 3x DYKDDDDK Peptide
Tricosa-peptide for competitively eluting DYKDDDK-tagged fusion proteins from immobilized supports
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
A368055 mg
A368065 x 5 mg
Número do catálogo A36805
Preço (BRL)
3.515,48
Each
Quantity:
5 mg
Preço (BRL)
3.515,48
Each
Pierce 3X DYKDDDDK Peptide is a lyophilized peptide with the sequence Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys(MDYKDHDGDYKDHDIDYKDDDDK). It has a molecular weight of 2861.9 g/mol. For use, reconstitute to 1.5 mg/mL in PBS.
This tricosa-peptide can be used to competitively elute DYKDDDDK-tagged fusion proteins from immobilized anti-DYKDDDDK affinity supports, especially when a DYKDDDDK-tagged protein is sensitive to low pH. At a 1.5 mg/mL concentration, complete release of tagged protein can be achieved in 5–10 minutes at ambient temperature.
Note: Pierce 3X DYKDDDDK Peptide may interfere with protein determination assays and absorbance at 280 nm. However, the peptide can be removed from the eluate by desalting (e.g., with Zeba Spin Desalting Columns, Cat. No. 89891).
For Research Use Only. Not for use in diagnostic procedures.
Especificações
For Use With (Application)Protein Purification and Isolation
FormLyophilized Powder
FormatLyophilized Powder
Product LinePierce
Product TypePeptides
Protein FormRecombinant
Quantity5 mg
Shipping ConditionWet Ice
MatrixAffinity Resin
Unit SizeEach
Conteúdo e armazenamento
5 mg lyophilized peptide. Store at -5 to -30°C.