Sanger Sequencing Kit
Applied Biosystems™

Sanger Sequencing Kit

The Applied Biosystems Sanger Sequencing Kit includes ExoSAP-IT Express reagent, a BigDye Terminator v3.1 Cycle Sequencing Kit, and a BigDye深入閱讀
Have Questions?
產品號碼Quantity
A38073200 Reactions
產品號碼 A38073
價格 (TWD)
92,500.00
Each
Quantity:
200 Reactions
價格 (TWD)
92,500.00
Each
The Applied Biosystems Sanger Sequencing Kit includes ExoSAP-IT Express reagent, a BigDye Terminator v3.1 Cycle Sequencing Kit, and a BigDye XTerminator Purification Kit, providing all the reagents needed to clean up PCR products, perform cycle sequencing, and clean up sequencing products. This simple, fast workflow enables completion of Sanger sequencing, from sample to answer, in less than a workday.

ExoSAP-IT Express reagent: fastest PCR cleanup method
In only 5 minutes, ExoSAP-IT Express PCR Product Cleanup Reagent removes excess primers and unincorporated nucleotides from a PCR reaction. Eliminate spin columns, magnetic beads, sedimentation, filtration, and gel purification. The novel ExoSAP-IT technology allows for a significant reduction in sample cleanup time with minimal steps, providing the simplest workflow.

• 5-min protocol
• One-tube, one-step PCR cleanup
• Novel enzyme technology
• 100% recovery of PCR products

BigDye Terminator v3.1 Cycle Sequencing Kit
The BigDye Terminator v3.1 Cycle Sequencing Kit provides premixed reagents for Sanger sequencing reactions. The kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.

• Improve the quality of your results for a wide range of sequencing applications
• Optimized for long read lengths
• Better dye mobility characteristics
• Improved performance reading through GT-rich regions
• Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
• Enhance your productivity and reduce costs

BigDye XTerminator Purification Kit
The BigDye XTerminator Purification Kit is a fast, simple purification method for DNA sequencing reactions that removes unincorporated BigDye terminators and salts. No more dye blobs! Cleanup is complete in under 40 minutes and requires less than 10 minutes of labor.

• Eliminate liquid transfer—use a single plate
• Obtain complete dye blob removal
• Stabilize samples before analysis
• Improve your sequencing workflow
• Perform manual or automated protocols
• Get rapid, reliable, and reproducible results

For Research Use Only. Not for use in diagnostic procedures.
規格
For Use With (Application)Standard Sequencing
For Use With (Equipment)Sequencing Instruments (Capillary Electrophoresis-Based)
No. of Reactions200
Product TypeSanger Sequencing Kit
Quantity200 Reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
FormatKit
Unit SizeEach
內容物與存放
• 2 x 100 rxns ExoSAP-IT Express, store at -5 to -30°C
• 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), store at 2–8°C
• 1 x 800 µL BigDye Terminator v3.1 ready reaction mix, store at -15 to -25°C
• 1 tube M13 (-21) primer, store at -15 to -25°C
• 1 tube pGEM control DNA, store at -15 to -25°C
• 1 x 2 mL XTerminator solution, store at 2–8°C
• 1 x 9 mL SAM solution, store at room temperature or 4°C

常見問答集 (常見問題)

What sequencing reaction volume do you recommend when using the Sanger Sequencing Kit?

When using the Sanger Sequencing Kit, we recommend using a sequencing reaction volume of 10 µL.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

For the Sanger Sequencing Kit, what is the recommended volume of purified PCR product in the sequencing reaction?

For the sequencing reaction, we recommend using 3 µL of purified PCR product (purified using ExoSAP-IT Express PCR Product Cleanup Reagent from the kit).

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What are the components of the Sanger Sequencing Kit, and what is the function of each component?

The Sanger Sequencing Kit includes the following components:
- ExoSAP-IT Express PCR Product Cleanup Reagent, which removes excess primers and unincorporated nucleotides from a PCR reaction
- BigDye Terminator v3.1 Cycle Sequencing Kit, which provides premixed reagents for Sanger sequencing reactions
- BigDye XTerminator Purification Kit, a fast, simple purification method for DNA sequencing reactions, that removes unincorporated BigDye terminators and salts

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Are the components in the Sanger Sequencing Kit available separately?

Yes, all the components in the Sanger Sequencing Kit are available separately. The individual components can be found on the following product pages:
- For the ExoSAP-IT Express PCR Product Cleanup Reagent please go here (https://www.thermofisher.com/order/catalog/product/75001.4X.1.ML). We recommend replacing the strip septa after each run. Bleach, detergents, or solvents should not be used to clean the septa. Use of these reagents can cause them to dry out faster and risk contaminating the system.
- For the BigDye Terminator v3.1 Cycle Sequencing Kit, please go here (https://www.thermofisher.com/order/catalog/product/4337454).
- For the BigDye XTerminator Purification Kit, please go here (https://www.thermofisher.com/order/catalog/product/4376486).

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What is the Sanger Sequencing Kit?

The Sanger Sequencing Kit is designed for our complete sequencing workflow on the SeqStudio, 3500 series, and 3730 series genetic analyzers. The new kit provides all the components needed to clean up a PCR product for sequencing, perform cycle sequencing, and then purify the sequencing product. This simple, fast workflow enables completion of Sanger sequencing, from sample to answer, in less than a workday.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.